missing translation for 'onlineSavingsMsg'
Learn More

Bisacodyl, TRC

Product Code. 30134423
Change view
Click to view available options
Quantity:
50 mg
1 g
10 g
Unit Size:
10g
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134423 50 mg 50mg
17951155 10 g 10g
17901805 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134423 Supplier Toronto Research Chemicals Supplier No. TRCB40080050MG
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 603-50-9
Molecular Formula C22 H19 N O4
Molecular Weight (g/mol) 361.39
Synonym Phenol, 4,4'-(2-pyridinylmethylene)bis-, diacetate (ester) (9CI), Phenol, 4,4'-(2-pyridylmethylene)di-, diacetate (6CI,7CI), Phenol, 4,4'-(2-pyridylmethylene)di-, diacetate (ester) (8CI), 4,4'-(2-Pyridylmethylene)diphenol diacetate, Alophen, Bicol, Bis(p-acetoxyphenyl)-2-pyridylmethane, Bisacodyl, Brocalax, Broxalax, Contralax, Correctol Caplets, Correctol Tablets, DAMP, Dulcolan, Dulcolax, Durolax, Endokolat, Eulaxan, Feen-a-Mint Tablets, Fenilaxan, Fleet, Godalax, Hillcolax, Ivilax, LA96a, Laco, Laxadin, Laxanin N, Laxans, Laxine, Laxorex, NSC 614826, Neolax, Nigalax, Perilax, Prepacol, Pyrilax, SK-Bisacodyl, Sanvacual, Stadalax, Telemin, Theralax, Ulcol, Videx, Zetrax
IUPAC Name [4-[(4-acetyloxyphenyl)-pyridin-2-ylmethyl]phenyl] acetate
SMILES CC(=O)Oc1ccc(cc1)C(c2ccc(OC(=O)C)cc2)c3ccccn3

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C22H19NO4/c1-15(24)26-19-10-6-17(7-11-19)22(21-5-3-4-14-23-21)18-8-12-20(13-9-18)27-16(2)25/h3-14,22H,1-2H3
Formula Weight 361.1314
Percent Purity >95%
Chemical Name or Material Bisacodyl
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.