missing translation for 'onlineSavingsMsg'
Learn More

Bisdemethoxycurcumin, TRC

Product Code. 30137535
Change view
Click to view available options
Quantity:
50 mg
500 mg
1 g
Unit Size:
1g
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137535 50 mg 50mg
30132790 1 g 1g
17967184 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137535 Supplier Toronto Research Chemicals Supplier No. TRCB42580550MG
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24939-16-0
Molecular Formula C19 H16 O4
Molecular Weight (g/mol) 308.33
Synonym Bisdesmethoxycurcumin
IUPAC Name 1,7-bis(4-hydroxyphenyl)hepta-1,6-diene-3,5-dione
SMILES Oc1ccc(C=CC(=O)CC(=O)C=Cc2ccc(O)cc2)cc1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C19H16O4/c20-16-7-1-14(2-8-16)5-11-18(22)13-19(23)12-6-15-3-9-17(21)10-4-15/h1-12,20-21H,13H2
Formula Weight 308.1049
Chemical Name or Material Bisdesmethoxycurcumin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.