missing translation for 'onlineSavingsMsg'
Learn More

Bismuth Subsalicylate, MedChemExpress

Product Code. 16490988
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16490988 5 g 5g
16408528 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16490988 Supplier Medchem Express Supplier No. HYB0550

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Bismuth Subsalicylate is a potent and orally active antacid and anti-diarrheal agent. Bismuth Subsalicylate reduces inflammation/irritation of stomach and intestinal lining through inhibition of prostaglandin synthesis in vivo. Bismuth Subsalicylate is widely used for the research of diarrheal disorders, including indigestion, diarrhoea, nausea, et al.

Bismuth Subsalicylate is a potent and orally active antacid and anti-diarrheal agent. Bismuth Subsalicylate reduces inflammation/irritation of stomach and intestinal lining through inhibition of prostaglandin synthesis in vivo. Bismuth Subsalicylate is widely used for the research of diarrheal disorders, including indigestion, diarrhoea, nausea, et al.

Specifications

Chemical Name or Material Bismuth Subsalicylate
CAS 14882-18-9
Color White
Purity Grade Notes Research
Molecular Formula C7H5BiO4
Quantity 5 g
Synonym Bismuth oxysalicylate Bismuth(III) salicylate basic
Solubility Information DMSO : 1 mg/mL (2.76 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.28 mM; Need ultrasonic)
SMILES O=C1O[Bi](O)OC2=CC=CC=C12
Molecular Weight (g/mol) 362.09
Formula Weight 362.09
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.