missing translation for 'onlineSavingsMsg'
Learn More

Bisphenol F, TRC

Product Code. 30006767
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006767 25 g 25g
30136014 5 g 5g
30008966 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006767 Supplier Toronto Research Chemicals Supplier No. TRCB51955525G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 620-92-8
Molecular Formula C13 H12 O2
Molecular Weight (g/mol) 200.23
Synonym Phenol, 4,4'-methylenedi- (8CI), Phenol, p,p'-methylenedi- (4CI), 4,4'-Methylenebis[phenol], 1,1-Bis(4-hydroxyphenyl)methane, 4,4'-Bis(hydroxyphenyl)methane, 4,4'-Dihydroxydiphenylmethane, 4,4'-Methylenediphenol, Bis(4-hydroxyphenyl)methane, Bis(p-hydroxyphenyl)methane, Bisphenol F, HDM, NSC 401136, PP-BIP-F, p,p'-BPF, p-(p-Hydroxybenzyl)phenol
IUPAC Name 4-[(4-hydroxyphenyl)methyl]phenol
SMILES Oc1ccc(Cc2ccc(O)cc2)cc1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C13H12O2/c14-12-5-1-10(2-6-12)9-11-3-7-13(15)8-4-11/h1-8,14-15H,9H2
Formula Weight 200.0837
Percent Purity >95%
Chemical Name or Material Bisphenol F
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.