missing translation for 'onlineSavingsMsg'
Learn More

Bitoscanate, MedChemExpress

Product Code. 16488646
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488646 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16488646 Supplier Medchem Express Supplier No. HYB1160
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Bitoscanate (p-Phenylene diisothiocyanate) is an organic chemical compound used in the treatment of hookworms.

Bitoscanate (p-Phenylene diisothiocyanate) is an organic chemical compound used in the treatment of hookworms.

Specifications

Chemical Name or Material Bitoscanate
Physical Form Solid
CAS 4044-65-9
Color Light Gray
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C8H4N2S2
For Use With (Application) COVID-19-immunoregulation
Synonym p-Phenylene diisothiocyanate 1,4-Diisothiocyanatobenzene PDITC
Solubility Information DMSO : 20 mg/mL (104.03 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES S=C=NC1=CC=C(N=C=S)C=C1
Molecular Weight (g/mol) 192.26
Formula Weight 192.26
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.