Learn More
Blasticidin S HCl, Corning™
For Research Use Only. Not Intended for Diagnostic or Therapeutic Use.
Brand: Corning Media Tech 30-100-RB
Code : NEW
Additional Details : CAS Number : 3513-03-9 Weight : 1.00000kg Transport : UN number : 2811 Chem class : 6.1 Pack group : II
Description
- CAS No: 3513-03-9
- Formula: C17H26N8O5 •HCl
- Pure substance/mixture: Substance
Specifications
Blasticidin S HCl | |
White | |
Powder | |
Antibiotic |