missing translation for 'onlineSavingsMsg'
Learn More

Blasticidin S Hydrochloride (From Streptomyces Griseochromogenes), Fisher BioReagents

Product Code. 10264913
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Packaging:
Amber Glass
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10264913 25 mg Amber Glass 25mg
10658203 50 mg Amber Glass 50mg
10648203 100 mg Amber Glass 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10264913 Supplier Fisher BioReagents Supplier No. BP264725
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 3-9-3513
Molecular Formula C17H27ClN8O5
Molecular Weight (g/mol) 458.90
InChI Key YQXYQOXRCNEATG-UHFFFAOYNA-N
Synonym BLA-S, Cytovirin
PubChem CID 124080960
IUPAC Name (2R,3S,6S)-3-[[(3R)-3-amino-5-[carbamimidoyl(methyl)amino]pentanoyl]amino]-6-(4-amino-2-oxopyrimidin-1-yl)-3,6-dihydro-2H-pyran-2-carboxylic acid;hydrochloride
SMILES CN(CCC(CC(=O)NC1C=CC(OC1C(=O)O)N2C=CC(=NC2=O)N)N)C(=N)N.Cl

Specifications

Type Blasticidin S Hydrochloride
Physical Form Powder/Solid
Melting Point 248°C
Packaging Amber Glass
Potency 900μg/mg min.
Color White
Quantity 25 mg
Assay Percent Range ≥99.0 %
Formula Weight 458.69
Percent Purity 99.0% min.
Chemical Name or Material Blasticidin S Hydrochloride, From Streptomyces griseochromogenes
Show More Show Less
compliance-icons
Product Identifier
  • Blasticidin S Hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.