Learn More
Description
Blasticidin S hydrochloride is a broad spectrum nucleoside antibiotic that inhibits protein synthesis in prokaryotic and eukaryotic organisms. It is suitable for selection of cells carrying plasmids conferring blasticidin resistance. Blasticidin S is used as a selection agent for transformed cells that contain the resistance genes bls, bsr, or BSD, has been used to select HEK293-T cells with TLR-2 constructs and HEK-D5 cells, and is also used to study protein synthesis at the level of peptide bond formation. Usage Statement: Unless specified otherwise, MP Biomedical™ products are for research or further manufacturing use only, not for direct human use.
Specifications
Specifications
| Physical Form | Powder |
| CAS | 3-9-3513 |
| Source | Streptomyces griseochromogenes |
| Sterility | Non-sterile |
| Quantity | 250 mg |
Safety and Handling
- Blasticidin S Hydrochloride
- Danger
- Acute toxicity Category 1
- H300-Fatal if swallowed.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: Blasticidin S Hydrochloride
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.