missing translation for 'onlineSavingsMsg'
Learn More

(-)-Blebbistatin, MedChemExpress

Product Code. 16488377
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488377 5 mg 5mg
16445688 10 mg 10mg
16401857 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16488377 Supplier Medchem Express Supplier No. HY13441
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress (-)-Blebbistatin is a selective inhibitor of the ATPase activity of non-muscle myosin II.

(-)-Blebbistatin is a selective inhibitor of the ATPase activity of non-muscle myosin II.

Specifications

Chemical Name or Material (-)-Blebbistatin
Physical Form Powder
CAS 856925-71-8
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C18H16N2O2
For Use With (Application) COVID-19-immunoregulation
Synonym (S)-(-)-Blebbistatin
Solubility Information DMSO : 10 mg/mL (34.21 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CC1=CC=C(N=C(N(C2=CC=CC=C2)CC3)[C@]3(O)C4=O)C4=C1
Molecular Weight (g/mol) 292.33
Formula Weight 292.33
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.