missing translation for 'onlineSavingsMsg'
Learn More

TOKU-E™ Bleomycin ReadyMade™ Solution

Product Code. 30044055
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 1 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30044055 10 x 1 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30044055 Supplier TOKU-E™ Supplier No. B09610x1ML
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Bleomycin ReadyMade™ Solution is provided as a sterile-filtered solution of Bleomycin Sulfate in water.

  • CAS Number: 9041-93-4
  • Molecular Formula: C55H84N17O21S3 (Bleomycin A2); C55H84N20O21S2 (Bleomycin B2)
  • Molecular Weight: 2927.17 g/mol
  • Storage Conditions: -20°C
  • Form: Liquid
  • Source: Streptomyces verticillus
  • Spectrum: Bleomycin is used as a selective agent for Gram-negative and Gram-positive bacteria
  • Mechanism of Action: Bleomycins act by intercalation of DNA and RNA. In the presence of oxygen and metal ions, notably copper and iron, Bleomycins form a pseudo-enzyme that induces DNA cleavage.

Bleomycin's anticancer activities include the increase of caspase-3 and p53, and the inhibition of telomerase activity leading to apoptosis. The anti-cancer properties derive from its ability to effect DNA cleavage in cancer cells.

Specifications

CAS 9041-93-4
Certifications/Compliance Purity >95%
Chemical Name or Material Bleomycin
Color Colorless
Content And Storage -20°C, Protect from light
Description Bleomycin ReadyMade™ Solution 10 mg/mL in Water
Concentration 10 to 10.5 mg/mL
Form Liquid
Product Type Antibiotic
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives None
Without Additives None
Formulation Liquid, in water
For Use With (Application) Microbiology, Plant Biology, Eukaryotic Cell Culture, Cancer Application
Product Line ReadyMade™ Solutions
Purity or Quality Grade >95%
Quantity 10 x 1 mL
Shipping Condition Ice Pack
Source Streptomyces verticillus
Species Bacteria
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.