missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BLMH/Bleomycin Hydrolase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98334-100ul
This item is not returnable.
View return policy
Description
BLMH/Bleomycin Hydrolase Polyclonal antibody specifically detects BLMH/Bleomycin Hydrolase in Mouse samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
| BLMH/Bleomycin Hydrolase | |
| Polyclonal | |
| Western Blot 1:500-1:1000, ELISA 1:25000-1:50000, Immunoprecipitation 0.2-1 μL/mg of lysate | |
| BHBMHBLM hydrolase, bleomycin hydrolase, EC 3.4.22.40 | |
| Produced in rabbits immunized with purified, recombinant Mouse BLMH/Bleomycin Hydrolase (Accession#: NP_848760.1; Asn 2-Glu 455) | |
| 100 μg | |
| Cancer, DNA Repair, Genes Sensitive to DNA Damaging Agents | |
| 642 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction