missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BLNK Recombinant Rabbit Monoclonal Antibody (K01_1N58)

Product Code. 30830431
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830431 50 μL
30829936 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830431 Supplier Invitrogen™ Supplier No. MA564668

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

BLNK Recombinant Monoclonal Antibody for Western Blot

B cell linker protein (Blnk) is a protein encoded by the Blnk gene located on chromosome 10 in humans. It is also known as SLP 65 (Src homology 2 domain-containing leukocyte protein of 65 kDa). It derives its name from being an adaptor protein in the B cell receptor signaling cascade with several supporting studies showing improper development of B cells in the absence of Blnk. Blnk undergoes tyrosine phosphorylation by Syk, a B cell receptor activated kinase, which provides a platform for interaction of signaling intermediaries such as the Vav GEF and Grb2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BLNK
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1N58
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 150mM NaCl, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene BLNK
Gene Accession No. Q4KM52, Q8WV28, Q9QUN3
Gene Alias AGM4; B cell adaptor containing SH2 domain; B cell linker; B cell linker protein; BASH; bca; B-cell activation; B-cell adapter containing a SH2 domain protein; B-cell adapter containing a Src homology 2 domain protein; B-cell linker; B-cell linker protein; Blnk; BLNK-S; Cytoplasmic adapter protein; LY57; Ly-57; lymphocyte antigen 57; Lyw-57; MGC111051; SLP65; Slp-65; Src homology [SH2] domain-containing leukocyte protein of 65 kD; Src homology 2 domain-containing leukocyte protein of 65 kDa
Gene Symbols BLNK
Host Species Rabbit
Immunogen A synthetic peptide of human BLNK.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17060, 29760, 499356
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.