missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Blooms Syndrome Recombinant Rabbit Monoclonal Antibody (HL2771)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556612
This item is not returnable.
View return policy
Description
Blooms Syndrome Recombinant Monoclonal Antibody for Western Blot
The protein encoded by this gene belongs to the annexin family of calcium-dependent phospholipid binding proteins some of which have been implicated in membrane-related events along exocytotic and endocytotic pathways. Annexin 5 is a phospholipase A2 and protein kinase C inhibitory protein with calcium channel activity and a potential role in cellular signal transduction, inflammation, growth and differentiation. Annexin 5 has also been described as placental anticoagulant protein I, vascular anticoagulant-alpha, endonexin II, lipocortin V, placental protein 4 and anchorin CII. The gene spans 29 kb containing 13 exons, and encodes a single transcript of approximately 1.6 kb and a protein product with a molecular weight of about 35 kDa.
Specifications
| Blooms Syndrome | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P54132 | |
| BLM | |
| Recombinant fragment of human BLM. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| HL2771 | |
| Unconjugated | |
| BLM | |
| Blm; Bloom syndrome homolog; Bloom syndrome protein; Bloom syndrome protein homolog; Bloom syndrome RecQ like helicase; Bloom syndrome, RecQ helicase-like; BS; DNA helicase, RecQ-like type 2; mBLM; MGC126616; MGC131618; MGC131620; RecQ helicase homolog; recQ protein-like 3; RecQ2; RECQL2; RECQL3 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 641 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction