missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMAL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
256.00 € - 627.00 €
Specifications
| Antigen | BMAL1 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 0.5 ug/mL - 2 ug/mL, Flow Cytometry, Immunohistochemistry 1:250. Use reported in scientific literature (PMID 33510438), Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:250, Immunohistochemistry-Frozen reported in scientific literature (PMID 23736292), Flow (Intracellular), Chromatin Immunoprecipitation (ChIP), Knockout Validated |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunofluorescence |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18739853
|
Novus Biologicals
NB100-2288SS |
0.025 mL |
256.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18720634
|
Novus Biologicals
NB100-2288 |
627.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
BMAL1 Polyclonal specifically detects BMAL1 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular), Knockout Validated.Specifications
| BMAL1 | |
| Western Blot 0.5 ug/mL - 2 ug/mL, Flow Cytometry, Immunohistochemistry 1:250. Use reported in scientific literature (PMID 33510438), Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:250, Immunohistochemistry-Frozen reported in scientific literature (PMID 23736292), Flow (Intracellular), Chromatin Immunoprecipitation (ChIP), Knockout Validated | |
| Polyclonal | |
| Rabbit | |
| Cholesterol Metabolism, Chromatin Research, Circadian Rhythm, Hypoxia, Lipid and Metabolism, Neuroscience, Transcription Factors and Regulators, Vision | |
| PBS with 0.02% Sodium Azide | |
| aryl hydrocarbon receptor nuclear translocator-like, aryl hydrocarbon receptor nuclear translocator-like protein 1, basic-helix-loop-helix-PAS orphan MOP3, BHLHE5, bHLHe5brain and muscle, bHLH-PAS protein JAP3, BMAL1TIC, Brain and muscle ARNT-like 1, Class E basic helix-loop-helix protein 5, Member of PAS protein 3, member of PAS superfamily 3, MOP3BMAL1c, PAS domain-containing protein 3, PASD3MGC47515 | |
| ARNTL | |
| IgG | |
| Affinity Purified | |
| 70 kDa |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Primate | |
| O00327 | |
| 406 | |
| Bacterially expressed human BMAL1 (C-terminus). [UniProt# O00327]. | |
| Primary | |
| Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title