missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMP-2 Rabbit anti-Human, Clone: 110, Novus Biologicals™
Rabbit Monoclonal Antibody
367.00 € - 552.00 €
Specifications
| Antigen | BMP-2 |
|---|---|
| Clone | 110 |
| Dilution | Immunocytochemistry/Immunofluorescence 1:20-1:100 |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18288396
|
Novus Biologicals
NBP2-89490-50UL |
50 μg |
367.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18245968
|
Novus Biologicals
NBP2-89490-100UL |
100 μg |
552.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BMP-2 Monoclonal specifically detects BMP-2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| BMP-2 | |
| Immunocytochemistry/Immunofluorescence 1:20-1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BMP-2, BMP-2A, BMP2ABone morphogenetic protein 2A, bone morphogenetic protein 2 | |
| Recombinant Human BMP-2 protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 110 | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Biologically Active Proteins, Cytokine Research, Immunology, Neuronal Cell Markers, Stem Cell Markers, Stem Cell Signaling Pathway | |
| 0.2 um filtered solution in PBS | |
| 650 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title