missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMP-9 Antibody (16S11) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
195.00 € - 510.00 €
Specifications
| Antigen | BMP-9 |
|---|---|
| Clone | 16S11 |
| Concentration | LYOPH |
| Applications | Blocking Assay, ELISA, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18130923
|
Novus Biologicals
NBP2-22034-0.025MG |
0.025 mg |
195.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18787053
|
Novus Biologicals
NBP2-22034 |
510.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
BMP-9 Monoclonal specifically detects BMP-9 in Human samples. It is validated for Western Blot, ELISA, Block/Neutralize.Specifications
| BMP-9 | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BMP9BMP-9Bone morphogenetic protein 9, GDF-2, growth differentiation factor 2, growth/differentiation factor 2 | |
| GDF2 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 16S11 | |
| Blocking Assay, ELISA, Western Blot | |
| Unconjugated | |
| Mouse | |
| Cytokine Research | |
| Q9UK05 | |
| 2658 | |
| IgG2 | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title