missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BMP5 Monoclonal Antibody (149107)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561768
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
TGF-β family members are key modulators of cell proliferation, differentiation, matrix synthesis, and apoptosis. As implied by their name, BMPs initiate, promote, and regulate the development, growth, and remodeling of bone and cartilage. In addition to this role, BMPs are also involved in prenatal development and postnatal growth, remodeling, and maintenance of a variety of other tissues and organs. BMP-5 is expressed in the nervous system, lungs and liver. It is a known regulator for dendritic growth in sympathetic neurons. BMP-5 is a 454 amino acid precursor protein that is cleaved to release the biologically active C-terminal mature protein.
Specifications
| BMP5 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P22003 | |
| Bmp5 | |
| Mouse myeloma cell line NS0-derived recombinant human BMP-5, Asn313-His454. | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Neutralization | |
| 149107 | |
| Unconjugated | |
| Bmp5 | |
| AU023399; BMP; BMP 5; Bmp5; BMP-5; bone morphogenenic protein-5; Bone morphogenetic protein; bone morphogenetic protein 5; MGC34244; se; short ear | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 653 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction