Learn More
Invitrogen™ BMS-936559/MDX-1105 Recombinant Monoclonal Antibody
GREENER_CHOICE
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA558375
 View more versions of this product
Product Code. 30283003
  • 477.00 € / 100µg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
BMS-936559/MDX-1105 is an anti-PD-L1 monoclonal antibody that blocks the interaction between PD-L1 and its receptors, PD-1 and CD80, thereby enhancing T-cell responses and promoting anti-tumor activity.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BMS-936559/MDX-1105
Recombinant Monoclonal
Unconjugated
MDX-1105
Protein A
RUO
Human
Antibody
IgG4SP
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized