missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BNIP1 Antibody (SR1417), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21748-50ul
This item is not returnable.
View return policy
Description
BNIP1 Monoclonal antibody specifically detects BNIP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| BNIP1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| SR1417 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| BCL2/adenovirus E1B 19 kDa protein-interacting protein 1, BCL2/adenovirus E1B 19kDa interacting protein 1, BCL2/adenovirus E1B 19kD-interacting protein 1, NIP1, SEC20, SEC20L, Transformation-related gene 8 protein, TRG-8, vesicle transport protein SEC20 | |
| A synthesized peptide derived from human BNIP1 (Uniprot #: Q12981) | |
| 50 μg | |
| Apoptosis | |
| 662 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction