missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BNIP3 Antibody (S09-1G8), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19550-100ul
This item is not returnable.
View return policy
Description
BNIP3 Monoclonal antibody specifically detects BNIP3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| BNIP3 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence | |
| S09-1G8 | |
| Unconjugated | |
| BCL2/adenovirus E1B 19 kDa protein-interacting protein 3, BCL2/adenovirus E1B 19kDa interacting protein 3, BCL2/adenovirus E1B 19kD-interacting protein 3, NIP3 | |
| A synthetic peptide of human BNIP3 (Uniprot # Q12983) | |
| 100 μg | |
| Apoptosis, Autophagy, Cancer, Hypoxia, Phospho Specific, Tumor Suppressors | |
| 664 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction