missing translation for 'onlineSavingsMsg'
Learn More

BOB.1 (B-Cell Marker) Mouse, Clone: BOB1/2424, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 5450-MSM4-P1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15990678

  • 485.00 € / 100µg
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

BOB.1 (B-Cell Marker)
Nuclear
BOB1/2424
Unconjugated
Mouse
35kDa
Protein A/G purified from Bioreactor concentrate.
RUO
5450
IgG2b κ
Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide.
Recombinant human BOB1 (POU2AF1) protein fragment (apprx. aa 148-255)
Ramos cells. Spleen or Tonsil
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers