missing translation for 'onlineSavingsMsg'
Learn More

Boc-Cys(Trt)-OH, TRC

Product Code. 30136961
Change view
Click to view available options
Quantity:
500 mg
1 g
10 g
Unit Size:
10g
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136961 10 g 10g
30133257 1 g 1g
30003615 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136961 Supplier Toronto Research Chemicals Supplier No. TRCB64629310G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21947-98-8
Molecular Formula C27 H29 N O4 S
Molecular Weight (g/mol) 463.59
Synonym N-(tert-Butoxycarbonyl)-S-trityl-L-cysteine, N-Boc-S-trityl-L-cysteine, N-tert-Butoxycarbonyl-S-tritylcysteine, NSC 129903;
IUPAC Name 2-(tert-butoxycarbonylamino)-3-tritylsulfanyl-propanoic acid
SMILES CC(C)(C)OC(=O)NC(CSC(c1ccccc1)(c2ccccc2)c3ccccc3)C(=O)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C27H29NO4S/c1-26(2,3)32-25(31)28-23(24(29)30)19-33-27(20-13-7-4-8-14-20,21-15-9-5-10-16-21)22-17-11-6-12-18-22/h4-18,23H,19H2,1-3H3,(H,28,31)(H,29,30)
Formula Weight 463.1817
Chemical Name or Material Boc-Cys(Trt)-OH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.