missing translation for 'onlineSavingsMsg'
Learn More

BOC-L-Serine hydrate, 98%

Product Code. 10043280 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10043280 5 g Glass Bottle 5g
10329060 25 g Glass Bottle 25g
10711355 100 g Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10043280 Supplier Thermo Scientific Acros Supplier No. 275660050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3262-72-4 | C8H15NO5 | 205.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3262-72-4
Molecular Formula C8H15NO5
Molecular Weight (g/mol) 205.21
MDL Number MFCD00037243
InChI Key FHOAKXBXYSJBGX-MLHKIVSYNA-N
Synonym boc-ser-oh, n-boc-l-serine, boc-l-serine, n-tert-butoxycarbonyl-l-serine, n-boc-serine, s-n-boc-serine, n-tert-butoxycarbonyl serine, s-2-tert-butoxycarbonyl amino-3-hydroxypropanoic acid, n-t-butoxycarbonyl-l-serine, boc-l-ser
PubChem CID 98766
SMILES CC(C)(C)OC(=O)N[C@@H](CO)C(O)=O

Specifications

Melting Point 84.0°C to 89.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula HOCH2CH(COOH)NHCOOC(CH3)3·
Specific Rotation − 8.10
Water 7.5 to 8.0% (K.F.)
IUPAC Name (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-hydroxypropanoic acid
Quantity 5 g
Flash Point 186°C
Formula Weight 205.21
Percent Purity 98%
Grade Technical
Physical Form Powder
Chemical Name or Material BOC-L-Serine hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.