missing translation for 'onlineSavingsMsg'
Learn More

BOC-O-Benzyl-L-serine hydroxysuccinimide ester, 97%, Thermo Scientific™

Product Code. 11965551 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11965551 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11965551 Supplier Thermo Scientific Acros Supplier No. 440890050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 13650-73-2
Melting Point 107°C to 109°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C19H24N2O7
Quantity 5 g
Specific Rotation -16.4
Synonym boc-ser bzl-osu, n-tert-butoxycarbonyl-o-benzyl-l-serine n-succinimidyl ester, s-2,5-dioxopyrrolidin-1-yl 3-benzyloxy-2-tert-butoxycarbonyl amino propanoate, 2,5-dioxopyrrolidin-1-yl 2s-3-benzyloxy-2-tert-butoxycarbonyl amino propanoate, boc-ser bzl osu, n-boc-3-benzyloxy-ala-succinimidyl oh, boc-ser bzl-osu hplc, n-boc-o-benzyl-l-serine n-succinimidyl ester, 2,5-dioxopyrrolidin-1-yl o-benzyl-n-t-butoxycarbonyl-l-serinate
InChI Key NTFYOALQRQBBDG-AWEZNQCLSA-N
SMILES CC(C)(C)OC(=O)NC(COCC1=CC=CC=C1)C(=O)ON2C(=O)CCC2=O
IUPAC Name (2,5-dioxopyrrolidin-1-yl) (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylmethoxypropanoate
Molecular Weight (g/mol) 392.41
PubChem CID 12879025
Formula Weight 392.41
Percent Purity 97%
Physical Form Powder
Chemical Name or Material BOC-O-Benzyl-L-serine hydroxysuccinimide ester
Show More Show Less
Product Identifier
  • BOC-O-Benzyl-L-serine hydroxysuccinimide ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.