missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Borate, 0.5M buffer soln., pH 8.5

Product Code. 15413509
Change view
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15413509 250 mL 250mL
15423509 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Código de producto. 15413509 Proveedor Thermo Scientific Chemicals N.º de proveedor J60803.AK
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 1330-43-4 | B4Na2O7 | 201.21 g/mol

Borate buffered saline has been used to determine whether heat-killed, borate-buffered suspensions of Gordonia bronchialis, Rhodococcus coprophilus or Tsukamurella inchonensis can be used to treat canine flea allergy. Borate buffered saline has also been used to investigate the effects of lentivirus mediated siRNA targeting human metastasis suppressor 1 (MTSS1, MIM-B gene) gene on the invasive and metastatic potentials of hepatocellular carcinoma (HCC) MHCC97H cells. Borate buffered saline has been used to determine the effects of packaging solution osmolality and buffering agent on soft contact lens parameters. BBS has many uses because it is isotonic and has a strong bactericidal effect. It can be used to dilute substances and in coating procedures. Additives such as Tween 20 and milk powder can be used to add function as washing buffer or blocking buffer. Finfs uses in Protein-coating on microtiter plates, Blocking solution in blots together with Casein or BSA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Borate buffered saline has been used to determine whether heat-killed, borate-buffered suspensions of Gordonia bronchialis, Rhodococcus coprophilus or Tsukamurella inchonensis can be used to treat canine flea allergy. Borate buffered saline has also been used to investigate the effects of lentivirus mediated siRNA targeting human metastasis suppressor 1 (MTSS1, MIM-B gene) gene on the invasive and metastatic potentials of hepatocellular carcinoma (HCC) MHCC97H cells. Borate buffered saline has been used to determine the effects of packaging solution osmolality and buffering agent on soft contact lens parameters. BBS has many uses because it is isotonic and has a strong bactericidal effect. It can be used to dilute substances and in coating procedures. Additives such as Tween 20 and milk powder can be used to add function as washing buffer or blocking buffer. Finfs uses in Protein-coating on microtiter plates, Blocking solution in blots together with Casein or BSA.

Solubility
Soluble in water 26 g/L (20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 1330-43-4
Molecular Formula B4Na2O7
Molecular Weight (g/mol) 201.21
MDL Number MFCD00081185
InChI Key NXSVMNYRVBQCMO-UHFFFAOYSA-N
IUPAC Name disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
SMILES [Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2

Especificaciones

Concentration 0.5 M
pH 8.5
Physical Form Liquid
Quantity 250 mL
Solubility Information Soluble in water 26g/L (20°C).
Chemical Name or Material Borate buffer soln.
compliance-icons
Nombre del producto
  • Borate, 0.5M buffer solution, pH 8.5
Palabra de advertencia
  • Danger
Clasificación
  • Reproductive toxicity Category 1B
Indicaciones de peligro
  • H360FD-May damage fertility. May damage the unborn child.
Consejos de prudencia
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Otros peligros
  • MIXTURE LIST-Contain: Boric acid

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.