missing translation for 'onlineSavingsMsg'
Learn More

Borax buffer solution, Honeywell Fluka™

Product Code. 15616940 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 L
Unit Size:
1 L
This item is not returnable. View return policy

Product Code. 15616940

Brand: Honeywell Fluka 734191L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

pH 10.00 (20°C)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 7732-18-5, 1330-43-4, 7732-18-5, 1310-73-2
Boiling Point 100°C (1.013 hPa)
CAS Min % ≥90.00%, ≥0.10%, ≥99.00%
CAS Max % ≤100.00%, <1.00%
Purity Grade Notes Ready-to-use buffers
Packaging Plastic Bottle
Quantity 1 L
Physical Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.