missing translation for 'onlineSavingsMsg'
Learn More

Boric acid, 98%

Product Code. 11437967
Change view
Click to view available options
Quantity:
1000 g
5000 g
Unit Size:
1000g
5000g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11437967 1000 g 1000g
11447967 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11437967 Supplier Thermo Scientific Alfa Aesar Supplier No. A16624.0B
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 10043-35-3 | BH3O3 | 61.83 g/mol

Boric Acid is a commonly used reagent in molecular biology for the preparation of buffers such as Tris Borate EDTA and has applications in capillary and gel electrophoresis. This compound has also been employed in a multitude of applictions, from weatherproofing fabrics to hardening steel. A commonly used molecular biology reagent that exhibits some antifungal activity

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Boric Acid is a commonly used reagent in molecular biology for the preparation of buffers such as Tris Borate EDTA and has applications in capillary and gel electrophoresis. This compound has also been employed in a multitude of applictions, from weatherproofing fabrics to hardening steel. A commonly used molecular biology reagent that exhibits some antifungal activity

Solubility
Soluble in glycerol, ether, alcohol, methanol, liquid ammonia, acetone (slightly), water (6.35 g/100 cc) at 30°C, water (27.6 g/100 cc) at 100°C, water (50 g/L) at 21°C, and water (1 g / 18 ml cold water).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Spécification

pH 3.8 to 4.8
Melting Point ∼185°C (decomposition)
Density 1.435
Odor Odorless
Flash Point None
Linear Formula H3BO3
Quantity 1000 g
Beilstein 1697939
Merck Index 14,1336
Solubility Information Soluble in glycerol,ether,alcohol,methanol,liquid ammonia,acetone (slightly),water (6.35g/100 cc) at 30°C,water (27.6g/100 cc) at 100°C,water (50g/L) at 21°C,and water (1 g / 18 ml cold water).
Formula Weight 61.83
Percent Purity 98%
Chemical Name or Material Boric acid
Afficher plus Afficher moins

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.