missing translation for 'onlineSavingsMsg'
Learn More

Boric acid, 99.99%, (trace metal basis), extra pure

Product Code. 10165850 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10165850 100 g Plastic bottle 100g
10299303 500 g Plastic bottle 500g
13438518 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Product Code. 10165850 Supplier Thermo Scientific Acros Supplier No. 315181000
  • ONLINE EXCLUSIVE PRICE
    79.95 €
    77.60 € / 100g
    Save 2.35 €
    2% Off
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price

CAS: 10043-35-3 | BH3O3 | 61.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis, Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

Color White
Melting Point 169.0°C
Packaging Plastic bottle
Assay Percent Range 99.99%
Quantity 100 g
Linear Formula H3BO3
Physical Form Crystals or Crystalline Powder
Fieser 01,63; 02,32; 03,29; 04,41; 05,48; 09,59; 11,70
Merck Index 15, 1339
Solubility Information Solubility in water: 49.5g/L (20°C). Other solubilities: 1g/18mL cold alcohol - 1g/6mL boiling alcohol, 1g/4mL glycerol, nacl, licl, mineral acids decrease the solubility, kno3,k2so4,kcl,nano3,na2so4 increase the sol.
Formula Weight 61.83
Percent Purity 99.99%
Grade Trace Metal Basis
Chemical Name or Material Boric acid, Extra Pure
Show More Show Less
compliance-icons
Product Identifier
  • Boric acid
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.