missing translation for 'onlineSavingsMsg'
Learn More

Boric Acid, for Electrophoresis, Fisher Chemical™

Product Code. 10754171 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Unit Size:
1kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10754171 5 kg Plastic keg 5kg
10496860 500 g Plastic powder jar 500g
10192180 1 kg Plastic powder jar 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10754171 Supplier Fisher Chemical Supplier No. B/P560/65
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 10043-35-3 | H3BO3

Boric Acid, for Electrophoresis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-35-3
Molecular Weight (g/mol) 61.83
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
IUPAC Name boric acid
SMILES OB(O)O

Specifications

pH 3.8 to 4.8
Color White
Melting Point 169°C
Packaging Plastic keg
Vapor Pressure 2.7mbar at 20°C
Quantity 5 kg
Molecular Formula BH3O3
Physical Form Powder Solid
MDL Number MFCD00011337
Formula Weight 61.83g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.