missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boric Acid, Granular, Ultrapure Bioreagent, J.T. Baker™
Shop All J.T.Baker Products
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Unit Size:
Each
Description
- For Molecular Biology Buffers
Specifications
Specifications
| Packaging | Poly Bottle |
| Specific Gravity | 1L = 1.43kg |
| Quantity | 500 g |
| Physical Form | Granular |
| Formula Weight | 61.83 |
| Grade | Ultrapure Bioreagent |
| Chemical Name or Material | Boric Acid |
Safety and Handling
Product Identifier
- Boric Acid
- Danger
- Reproductive toxicity Category 1B
- H360FD-May damage fertility. May damage the unborn child.
- P201-Obtain special instructions before use.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Boric acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction