missing translation for 'onlineSavingsMsg'
Learn More

Boric acid, Puratronic™, 99.9995% (metals basis)

Product Code. 11340879
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11340879

Brand: Thermo Scientific Alfa Aesar 010659.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10043-35-3 | BH3O3 | 61.83 g/mol

It is used as a primary or adjunct buffer in swimming pools, flame retardant, neutron absorber in nuclear power plants, colorant to make flame green, preservative in urine samples, welding flux in combination with borax, or precursor to other chemical compounds. It is used to prevent/remove existing wet and dry rot in timbers, treat wood against fungal/insect attack, prevent athlete’s foot, lubricate carom boards, and reduce surface oxidation on metals at high temperatures. It can be used to produce lubricant for ceramic or metal surfaces, textile fiberglass, induction furnace linings and ceramics, Silly Putty, and glass LCD flat panel displays. It can prevent the high temperature reaction of aluminum with nitrates in pyrotechniques, and neutralize active hydrofluoric acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

pH 3.8 to 4.8
Color White
Melting Point ∼185°C (decomposition)
Density 1.435 g/mL
Odor Odorless
Flash Point None
Assay Percent Range 99.99% (metals basis)
Quantity 500 g
Linear Formula H3BO3
Physical Form Powder
Beilstein 1697939
Merck Index 14,1336
Solubility Information Soluble in water- solubility increases rapidly with temperature. Soluble in methanol and ethanol. Very slightly soluble in acetone.
Formula Weight 61.83
Percent Purity 99.9995%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Boric acid
Visa mer Visa mindre

RUO – Research Use Only

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.