missing translation for 'onlineSavingsMsg'
Learn More

Boron solution for ICP/MS 10mg/L H2O matrix, Fisher chemical™
BIOPHARMA

Product Code. 12907333 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
50 mL
100 mL
Packaging:
Bottle
Unit Size:
100mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
12907333 100 mL Bottle 100mL
12997323 50 mL Bottle 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12907333 Supplier Fisher Chemical Supplier No. S 605 100
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula B

Specifications

Packaging Bottle
Concentration 10 mg/L
Solvent or Matrix Water
Quantity 100 mL
Chemical Name or Material Boron
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.