missing translation for 'onlineSavingsMsg'
Learn More

Boron Standard: B @ 1000 μg/mL in NH4OH, VHG™

Product Code. 30040680
Change view
Click to view available options
Quantity:
50 mL
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
50mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30040680 50 mL 50mL
30041315 500 mL 500mL
30041330 250 mL 250mL
30041328 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30040680 Supplier VHG Labs Supplier No. VHGPBZ50
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Boron Standard: B @ 1000 μg/mL in NH4OH
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Boron
CAS 7440-42-8
Methods ICP/ICP-MS
Analyte or Component Names Boron
Shipping Condition Room Temperature
Molecular Formula B
Certifications/Compliance ISO 17034
Synonym B
InChI Formula InChI=1S/B
SMILES [B]
IUPAC Name boron
Solvent or Matrix Ammonium Hydroxide
Molecular Weight (g/mol) 10.81
Quantity 50 mL
Formula Weight 11.009 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.