missing translation for 'onlineSavingsMsg'
Learn More

Boron Standard: B @ 2500 mg/Kg in 75 cSt oil

Product Code. 30327787
Change view
Click to view available options
Quantity:
400 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30327787 400 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30327787 Supplier VHG Labs Supplier No. VHGOB2500400G
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Boron
CAS 7440-42-8
Analyte or Component Names Boron
Shipping Condition Room Temperature
Molecular Formula B
Concentration 2500 mg/kg
Synonym B
InChI Formula InChI=1 S/B
Solution Type Mixture
SMILES [B]
Element B
IUPAC Name boron
Solvent or Matrix 75 cSt Oil
Industry Type Industrial QC
Molecular Weight (g/mol) 10.81
Quantity 400 g
Formula Weight 11.009
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.