missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Bottled Media for Sartorius Membrane Filtration Units

Product Code. 10544951 Shop All Sartorius Products
Change view
Click to view available options
For Use With (Application):
Membrane filtration method or for direct incubation, for determination of Escherichia coli and Coliforms
Membrane filtration method or for direct incubation, for determination of LactoBacilli and Pediococci and other beer spoiling organisms
Membrane filtration method or for direct incubation, for determination of Yeasts and molds
Membrane filtration method or for direct incubation, for determination of total count
Membrane filtration method or for direct incubation, for determination of wild Yeasts
Unit Size:
Pack of 4
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
10544951 Membrane filtration method or for direct incubation, for determination of total count Pack of 4
10440472 Membrane filtration method or for direct incubation, for determination of Yeasts and molds Pack of 4
11790875 Membrane filtration method or for direct incubation, for determination of wild Yeasts Pack of 4
10069600 Membrane filtration method or for direct incubation, for determination of Escherichia coli and Coliforms Pack of 4
10475401 Membrane filtration method or for direct incubation, for determination of LactoBacilli and Pediococci and other beer spoiling organisms Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10544951 Supplier Sartorius Supplier No. 14144A
  • ONLINE EXCLUSIVE PRICE
    174.00 €
    156.13 € / Pack of 4
    Save 17.87 €
    10% Off
View all promotions
 Restricted Item
Estimated Shipment: 11-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lysine agar media for wild yeasts determination

The traditional culture media for microorganisms is agar media. This can be used for the membrane filtration method or for direct incubation. Agar media in bottles are the cost-effective alternative for casting plates.

  • 250mL bottles, 4 bottles per box
  • Economical, provides time and cost savings
  • According to international formulas
  • Consistent quality
TRUSTED_SUSTAINABILITY

Specifications

Country of Origin Germany
Form Liquid
Format Bottle
Manufacturing Origin Germany
Media Type Nutrient
Product Type Culture Medium
Sterility Sterile
Test Type Standard Plate Count for Microorganism
Fill Volume 250 mL
For Use With (Application) Membrane filtration method or for direct incubation, for determination of total count
For Use With (Equipment) Sartorius Membrane Filtration Units
Packaging Type Bottle
Quantity 250 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.