missing translation for 'onlineSavingsMsg'
Learn More

BPTU, MedChemExpress

Product Code. 16448527
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16448527 5 mg 5mg
16436346 100 mg 100mg
16405888 10 mg 10mg
16403396 25 mg 25mg
16402037 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16448527 Supplier Medchem Express Supplier No. HY13831
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress BPTU (BMS-646786) is a non-nucleotide P2Y1 receptor allosteric antagonist with antithrombotic activity. BPTU is able to block the P2Y1 receptor located at the neuromuscular junction of the gastrointestinal tract.

BPTU (BMS-646786) is a non-nucleotide P2Y1 receptor allosteric antagonist with antithrombotic activity. BPTU is able to block the P2Y1 receptor located at the neuromuscular junction of the gastrointestinal tract.

Specifications

Chemical Name or Material BPTU
CAS 870544-59-5
Color White
Purity Grade Notes Research
Molecular Formula C23H22F3N3O3
Quantity 5 mg
Synonym BMS-646786
Solubility Information DMSO : ≥ 33.3 mg/mL (74.76 mM)
SMILES O=C(NC1=CC=C(OC(F)(F)F)C=C1)NC2=CC=CN=C2OC3=CC=CC=C3C(C)(C)C
Molecular Weight (g/mol) 445.43
Formula Weight 445.43
Percent Purity 99.84%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.