missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ BRANDplates cellGrade™ 384-Well, cellGrade-Treated, Flat-Bottom Microplate

Product Code. 15644017 Shop All Brand Products
Change view
Click to view available options
Packaging:
5/Pk., 32/Cs.
Unit Size:
Pack of 5
Product Code. Packaging unitSize
15644017 5/Pk., 32/Cs. Pack of 5
1 options
This item is not returnable. View return policy

Product Code. 15644017

Brand: BRAND™ 781974

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Color: White, Clear Bottom; Sterility: Sterile; Lid: With Lid

  • Hydrophilic surface with freely-accessible carboxyl and hydroxyl groups
  • Easily binds serum components to for indirect adhesion of cells
  • Sterilized by β-radiation to ISO 11137 and AAMI guidelines for a SAL of 10-6
  • Meet the requirements of Ph. Eur. and the USP 29
  • DNA-, DNase-, RNase-free
  • Endotoxins are <0.01 EU/mL
  • Orange alphanumeric embossing for easy well identification

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO 11137, DIN EN ISO 10993
Lid With Lid
Packaging 5/Pk., 32/Cs.
Color White, Clear Bottom
Material Polystyrene
No. of Wells 384
Sterility Sterile
Surface Treatment cellGrade
Well Shape Flat Bottom (F-well)
For Use With (Application) Cultivation of adherent cell lines
No. per Pack 5
Quantity 32/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.