missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ BRANDplates cellGrade™ 96-well PS Microplates

Product Code. 10532094 Shop All Brand Products
Change view
Click to view available options
Packaging:
1/Pk., 50/Cs.
5/Pk.
Color:
Black
Black, Clear Bottom, Clear Bottom
Clear
White
White, Clear Bottom, Clear Bottom
Well Shape:
Conical Bottom (V-well)
Flat Bottom (F-well)
Round Bottom (U-well)
Unit Size:
Pack of 5
Pack of 50
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Color Well Shape unitSize
10532094 1/Pk., 50/Cs. Black, Clear Bottom, Clear Bottom Flat Bottom (F-well) Pack of 50
15654017 5/Pk. Black, Clear Bottom, Clear Bottom Flat Bottom (F-well) Pack of 5
10307203 - Clear Round Bottom (U-well) Pack of 50
10002793 - Clear Conical Bottom (V-well) Pack of 50
10367243 - Clear Flat Bottom (F-well) Pack of 50
10617713 - White Flat Bottom (F-well) Pack of 50
10554584 - Black Flat Bottom (F-well) Pack of 50
10216523 - White, Clear Bottom, Clear Bottom Flat Bottom (F-well) Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 10532094 Supplier BRAND™ Supplier No. undefined
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Sterile 96-well microplates with cellGrade™surface treatment for optimal growth. Ideal for cultivating adherent cells with a hydrophilic PS surface featuring accessible carboxyl and hydroxyl groups. Enhances serum component binding for effective indirect cell adhesion.

  • Hydrophilic surface with freely-accessible carboxyl and hydroxyl groups
  • Easily binds serum components to for indirect adhesion of cells
  • Sterilized by B-radiation to ISO 11137 and AAMI guidelines for a SAL of 10-6
  • Meet the requirements of Ph. Eur. and the USP 29
  • BIO-CERT™ CELL CULTURE QUALITY means inserts: free of pyrogens, endotoxins, human DNA, RNase, and DNase
  • Endotoxins are <0.01 EU/mL
  • Orange alphanumeric embossing for easy well identification

Order Info

Packs of 50 individually wrapped plates with lids

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Certifications/Compliance ISO 11137, DIN EN ISO 10993
Format Standard
Lid With Lid
Array 8 x 12
Packaging 1/Pk., 50/Cs.
Binding Type Carboxyl and Hydroxyl Groups
Color Black, Clear Bottom, Clear Bottom
Product Type 96-well microplate
Material Polystyrene
Volume (Metric) Well 330 μL
No. of Wells 96
Sterility Sterile
Surface Treatment cellGrade
Total Volume (Metric) Well 330 μL
Well Shape Flat Bottom (F-well)
For Use With (Application) Cultivation of adherent cell lines
No. per Case 50/Cs.
No. per Pack 1/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.