Learn More
BRAND™ Deep Well Plate Mat Covers
Provide an effective plug seal for each well. Use of cover mats reduces the maximum volume of wells, but improves sealing and reduces evaporation of sample.
Brand: BRAND™ 701362
76.85 EUR valid until 2024-12-31
Use promo code "23264" to get your promotional price.
Code : LC
Additional Details : Weight : 0.54000kg
Description
- For use with 96-well plates, 364-well plates, and deep-well plates
- Polypropylene, polyethylene and silicone mats are autoclavable (EVA is not autoclavable.)
- Improves sealing and reduces evaporation of sample.
Specifications
96 | |
Cover Mat | |
Microplate, 2.2 mL |
24 | |
Square |