missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Deep Well Plates

Product Code. 10089910 Shop All Brand Products
Change view
Click to view available options
Volume (Metric) Well:
0.3 mL
0.5 mL
1 mL
1.1 mL
1.2 mL
2.2 mL
Format:
Elevated Wells
Standard
Well Shape:
Round
Round Bottom (U-well)
Square, Conical Bottom (V-well)
Square, Round Bottom (U-well)
Profile:
Elevated Rim
Low Profile
Standard
U-bottom
Unit Size:
1 pack
Pack of 24
Pack of 32
Pack of 48
Pack of 50
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Well Format Well Shape Profile Unit Size
10089910 2.2 mL Standard Square, Round Bottom (U-well) U-bottom Pack of 24
10680763 0.5 mL Standard Round Bottom (U-well) U-bottom Pack of 48
10060072 1.1 mL Standard Round Bottom (U-well) U-bottom Pack of 32
10538971 1.1 mL Standard Round U-bottom Pack of 24
17871242 1 mL Standard Square, Round Bottom (U-well) Low Profile Pack of 50
17881242 2.2 mL Standard Square, Conical Bottom (V-well) Standard Pack of 50
10688819 1.1 mL Standard Round Bottom (U-well) Low Profile Pack of 50
10493002 0.3 mL Standard Square, Conical Bottom (V-well) U-bottom Pack of 48
15613987 1.2 mL Elevated Wells Round Elevated Rim 1 pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 10089910 Supplier BRAND™ Supplier No. 701354
Discount Lab Supplies
Use promo code 28170
View all promotions
In Stock
Add to basket
Edge
Add to basket
Bulk Quote Request

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Seamlessly integrates into multi-channel systems and automation workflows.

BRAND™ Deep Well Plates are designed for a broad range of applications, including sample storage, high-throughput screening (HTS), cell and tissue culture, serial dilutions, and reagent transfers. Most plates are manufactured from high-quality virgin polypropylene, with one highly transparent polystyrene variant for applications requiring optical clarity.

  • Available in 96- and 384-well formats
  • Manufactured from high-quality virgin polypropylene or polystyrene
  • BIO-CERT™ PCR Quality are free from human DNA, DNase, RNase, and pyrogens
  • Polypropylene plates are autoclavable, chemically resistant, and suitable for storage down to -80°C (-112°F)
  • Polystyrene plate offers excellent transparency and is suitable for storage down to -20°C (-4°F)
  • ANSI/SLAS 1 and 4 compliant for compatibility with automated systems
  • Alphanumeric well labeling and stackable design for efficient organization
  • Notched corners ensure correct orientation in robotic and automated liquid-handling systems
  • Compatible with sealing mats or adhesive films to minimize evaporation and contamination
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Format Standard
Array 8 x 12
Autoclavable Autoclavable
Color Clear
Product Type Deep-well Plate
Material Polypropylene
Volume (Metric) Well 2.2 mL
No. of Wells 96
Sterility Non-sterile
Surface Treatment Untreated
Well Shape Square, Round Bottom (U-well)
For Use With (Application) Flexible mat covers or adhesive sealing films to reduce sample evaporation and contamination
No. per Pack 24
Profile U-bottom
Quantity 24 Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.