missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Real-Time PCR (qPCR) 96-Well Microplates, BIO-CERT™ PCR QUALITY, Low Profile

Product Code. 11668439 Shop All Brand Products
Click to view available options
Includes:
Plates and polyester seals
Plates only
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 11668439

Brand: BRAND™ 781364

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Optimized for accurate qPCR results, these plates are fully compatible with Roche™ LightCycler™ Systems.

Designed for optimal performance in fluorescence signal analysis and DNA quantification, these plates are ideal for qPCR applications. Key features include:

  • Enhanced Signal Reflection: White surface and titanium dioxide (TiO2) coloring maximize fluorescence signal reflection for precise quantification.
  • Exceptional Purity: BIO-CERT™ PCR QUALITY ensures plates are free of human DNA, RNase, DNase, pyrogens, and PCR inhibitors.
  • Advanced Manufacturing Standards: Produced in state-of-the-art clean room conditions for consistent quality.
  • Superior Thermal Performance: Extra-thin walls provide efficient thermal transfer.
  • User-Friendly Design:
    • Smooth surfaces optimize signal reflection
    • Blue embossed alphanumeric coding and a cut corner design ensure easy orientation
    • Low-profile, semi-skirted frame with a non-elevated rim offers compatibility with various thermal cyclers, including Roche™ LightCycler™ systems
  • Additional Convenience: Part no. 781365 includes 50 sheets of qPCR polyester sealing film
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Binding Property Untreated
Certifications/Compliance DNA, DNase, RNase-free
Format Standard
Array 8 x 12
Autoclavable Autoclavable
Packaging Bagged
Color White
Disposable Single-use
Product Type qPCR 96-well plate
For Use With (Equipment) Thermal Cyclers
Skirt Style Semi-skirted
Material Polypropylene
Volume (Metric) Well 0.15 mL
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Conical Bottom (V-well)
Dnase Free Yes
For Use With (Application) PCR
Includes Plates only
Profile Low
Quantity 50/Pk.
RNase Free Yes
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.