missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ BRANDplates™ pureGrade™ 1536-Well Microplates

Product Code. 10286703 Shop All Brand Products
Change view
Click to view available options
Color:
Black
Clear
White
Unit Size:
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10286703 Clear Pack of 50
10468423 White Pack of 50
10606953 Black Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10286703 Supplier BRAND™ Supplier No. 781640
Discount Lab Supplies
Use promo code 28170
View all promotions
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Non-treated microplates for diverse applications

  • Nontreated PS surface
  • Medium binding, nonsterile
  • Standard plate for most applications
  • Available in clear, white or black
  • Free from endotoxins, DNase, DNA, RNase; noncytotoxic

Homogenous assays, screening, and storage.

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Binding Property Medium Binding
Certifications/Compliance ISO 11137, AAMI
Format Standard
Lid Without Lid
Array 32 x 48
Autoclavable Not Autoclavable
Binding Type Untreated
Color Clear
Disposable Single-use
Product Type 1536-Well Cell Culture Multiwell Plates
For Use With (Equipment) Plate Readers
Skirt Style Full
Material Polystyrene
Volume (Metric) Well 10 μL
No. of Wells 1536
Volume (Metric) Working 10 μL
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Square, Flat Bottom (F-well)
For Use With (Application) Sterile applications, bacteriological assays
Quantity 50/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.