Learn More
BRAND™ inertGrade™ 96-well Microplates
Product Code. 15634017 Shop All Brand Products
Change view
Click to view available options
Color:
Black
White
Unit Size:
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15634017 Black Pack of 5
15624017 White Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15634017 Supplier BRAND™ Supplier No. 781913
  • ONLINE EXCLUSIVE PRICE
    137.00 €
    135.30 € / Pack of 5
    Save 1.70 €
    1% Off
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
Especially suited for cell cultures when adhesion is not desired.Reduction of cell adhesion and protein adsorption minimizes enzyme and cellular activation to aid in the inhibition of the differentiation of progenitor or stem cells and promotes the formation of spheroids.
  • Hydrogel technology optimizes the surface characteristics to suppress interactions of cells or protein with the polystyrene plastic.
  • Polystyrene construction
  • Sterile
  • 96 wells
  • Flat or Round Bottom
  • Black, White or Clear
  • Includes lid
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid With Lid
Color Black
Material Polystyrene
No. of Wells 96
Sterility Sterile
Well Shape Flat Bottom (F-well)
No. per Pack 5