missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAP Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92871-0.1ml
This item is not returnable.
View return policy
Description
BRAP Polyclonal antibody specifically detects BRAP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| BRAP | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| BRAP2RING finger protein 52, BRCA1 associated protein, EC 6.3.2.-, IMPBRCA1-associated protein, Impedes mitogenic signal propagation, Renal carcinoma antigen NY-REN-63, RNF52galectin-2-binding protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human BRAP (NP_006759.3). MSVSLVVIRLELAEHSPVPAGFGFSAAAGEMSDEEIKKTTLASAVACLEGKSPGEKVAIIHQHLGRREMTDVIIETMKSNPDELKTTVEERKSSEASPTAQRSKDHSKECINAAPDSPSKQLPDQISFFSGNPSVEIVHGIMHLYKTNKMTSLKEDVRRSAMLCILTVPAAMTSHDLMKFVAPFNEVIEQMKIIRDSTPN | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 8315 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction