missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRCA1 Antibody (KEN) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
200.00 € - 481.00 €
Specifications
| Antigen | BRCA1 |
|---|---|
| Clone | KEN |
| Concentration | 1.5 mg/mL |
| Dilution | Western Blot 2-4 ug/ml, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18041344
|
Novus Biologicals
NB100-599SS |
0.025 mL |
200.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18767103
|
Novus Biologicals
NB100-599 |
481.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
BRCA1 Monoclonal specifically detects BRCA1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| BRCA1 | |
| 1.5 mg/mL | |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Breast Cancer, Cancer, Cell Biology, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Tumor Suppressors, Ubiquitin Proteasome Pathway, Zinc Finger | |
| P38398 | |
| 672 | |
| Human BRCA1 corresponding to residues 1314-1864 [UniProt# P38398] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| KEN | |
| Western Blot 2-4 ug/ml, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BRCAI, breast and ovarian cancer susceptibility protein 1, breast and ovarian cancer sususceptibility protein, breast cancer 1, early onset, breast cancer type 1 susceptibility protein, EC 6.3.2, EC 6.3.2.-, IRIS, PNCA4, PSCP, subunit 1 | |
| BRCA1 | |
| IgG3 κ | |
| Protein A purified | |
| 240 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title