Learn More
Invitrogen™ BRCA2 Recombinant Rabbit Monoclonal Antibody (PSH07-55)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA560184
Description
Positive Control: HeLa cell lysate, 293T cell lysate, Jurkat cell lysate, MCF7. Tissue Specificity: Tissue enhanced (bone marrow, lymphoid tissue, testis). Subcellular Location: Nucleus, Cytoplasm, cytoskeleton, microtubule organizing center, centrosome. Sequence Similarities: 60% Mouse/Rat. Predicted band size: 384 kDa.
Breast Cancer Type 2 susceptibility protein (BRCA2) is a tumor-suppressor protein reported to be involved in double-strand break repair, homologous recombination and S-phase checkpoint activation.
Specifications
| BRCA2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P51587 | |
| BRCA2 | |
| Recombinant protein within human BRCA2 aa 3,219-3,418. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| PSH07-55 | |
| Unconjugated | |
| BRCA2 | |
| brca 2; BRCA1/BRCA2-containing complex, subunit 2; BRCA2; brca-2; BRCA2 DNA repair associated; BRCA2, DNA repair associated; BRCC2; breast and ovarian cancer susceptibility gene, early onset; breast and ovarian cancer susceptibility protein 2; breast cancer 2; breast cancer 2 tumor suppressor; breast cancer 2, early onset; breast cancer 2, mutation 1, University of Wisconsin-Madison; breast cancer susceptibility protein 2; breast cancer type 2 susceptibility protein; Breast cancer type 2 susceptibility protein homolog; BROVCA2; FACD; FAD; FAD1; FANCB; FANCD; FANCD1; Fanconi anemia group D1 protein; Fanconi anemia group D1 protein homolog; GLM3; mutant BRCA2; PNCA2; RAB163; RP11-298P3.4; XRCC11 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 675 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.