missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRD4 Antibody (RAB-C131) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
267.00 € - 624.00 €
Specifications
| Antigen | BRD4 |
|---|---|
| Clone | RAB-C131 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18642505
|
Novus Biologicals
NBP3-20125-0.025mg |
0.025 mg |
267.00 €
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18684834
|
Novus Biologicals
NBP3-20125-0.2mg |
0.2 mg |
624.00 €
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BRD4 Monoclonal antibody specifically detects BRD4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, ChIP assay (ChIP)Specifications
| BRD4 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| bromodomain containing 4, bromodomain-containing 4, CAPchromosome-associated protein, HUNK1bromodomain-containing protein 4, HUNKI, MCAP, Protein HUNK1 | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing BRD4 protein under non-denaturing conditions. | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
| RAB-C131 | |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cancer, Signal Transduction | |
| PBS | |
| 23476 | |
| IgG κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title