missing translation for 'onlineSavingsMsg'
Learn More

BRD7552, MedChemExpress

Product Code. 16467666
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16467666

Brand: Medchem Express HY19694

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress BRD7552, a potent PDX1 transcription factor inducer, upregulates PDX1 expression in both primary human islets and ductal cells, and induces epigenetic changes in the PDX1 promoter consistent with transcriptional activation. BRD7552 increases insulin expression. PDX1 is a key transcription factor involved in pancreas development and β cell function.

BRD7552, a potent PDX1 transcription factor inducer, upregulates PDX1 expression in both primary human islets and ductal cells, and induces epigenetic changes in the PDX1 promoter consistent with transcriptional activation. BRD7552 increases insulin expression. PDX1 is a key transcription factor involved in pancreas development and β cell function.

Specifications

Chemical Name or Material BRD7552
CAS 1137359-47-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C33H33N3O15
Quantity 100 mg
Solubility Information DMSO : ≥ 38 mg/mL (53.40 mM)
SMILES OC[C@@H](O1)[C@@H](OC(NC2=CC=C(C(OCC)=O)C=C2)=O)[C@H](OC(NC3=CC=C(OCO4)C4=C3)=O)[C@@H](OC(NC5=CC=C(OCO6)C6=C5)=O)[C@H]1OC
Molecular Weight (g/mol) 711.63
Formula Weight 711.63
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.