Learn More
Novus Biologicals™ Breast Matched Tumor and Normal Tissue Lysate
Product Code. 18165798 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
2 Vials
Unit Size:
Pack of 2
Product Code. Quantity Unit Size
18165798 2 Vials Pack of 2
1 options
Product Code. 18165798 Supplier Novus Biologicals™ Supplier No. NBP2471052VIAL
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Two matched 1 mg/mL vials (100 μg/vial) of human breast tissue lysate, one containing normal tissue and the other tumor tissue, for use in research. Lysates are guaranteed for 6 months from date of receipt.
The Breast Matched Tumor and Normal Tissue Lysate is derived from human.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 2 Vials
Content And Storage Store at -80C. Avoid freeze-thaw cycles.