missing translation for 'onlineSavingsMsg'
Learn More

Brexpiprazole, MedChemExpress

Product Code. 16406838
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16406838

Brand: Medchem Express HY15780

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Brexpiprazole (OPC-34712), an atypical orally active antipsychotic drug, is a partial agonist of human 5-HT1A and dopamine D2L receptor with Kis of 0.12 nM and 0.3 nM, respectively. Brexpiprazole is also a 5-HT2A receptor antagonist with a Ki of 0.47 nM. Brexpiprazole also shows potent antagonist activity at human noradrenergic α1B (Ki=0.17 nM) and α2C receptors (Ki=0.59 nM).

Brexpiprazole (OPC-34712), an atypical orally active antipsychotic drug, is a partial agonist of human 5-HT1A and dopamine D2L receptor with Kis of 0.12 nM and 0.3 nM, respectively. Brexpiprazole is also a 5-HT2A receptor antagonist with a Ki of 0.47 nM. Brexpiprazole also shows potent antagonist activity at human noradrenergic α1B (Ki=0.17 nM) and α2C receptors (Ki=0.59 nM).

Specifications

Chemical Name or Material Brexpiprazole
CAS 913611-97-9
Color White
Purity Grade Notes Research
Molecular Formula C25H27N3O2S
Quantity 10 mg
Synonym OPC-34712
Solubility Information DMSO : 25 mg/mL (57.66 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C1NC2=C(C=CC(OCCCCN3CCN(C4=C(C=CS5)C5=CC=C4)CC3)=C2)C=C1
Molecular Weight (g/mol) 433.57
Formula Weight 433.57
Percent Purity 98.25%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis